BRAND® Mortars and Pestles
Supplier: Brandtech
New Product
Certificates
About this item
Mortars and pestles made from a durable melamine formaldehyde (MF) resin, ideal for grinding and crushing a variety of materials.
- Autoclavable up to 121 °C
- Exceptional resistance to boiling and scratching
- Improved hardness, with a smooth surface texture
- Mortars come equipped with a foot and spout for added convenience
- Pestles are shaped for effortless gripping
Melamine formaldehyde resin, renowned for its thermal stability and remarkable resistance to various chemicals, makes BRAND® mortars and pestles ideal for grinding a diverse array of substances without the worry of corrosion or contamination.
The non-porous, smooth surface streamlines efficient grinding and effortless cleaning, while their durability ensures long-lasting performance.
Ordering information: Mortar and pestle are each sold separately.
Customers Who Bought This Also Bought