Order Entry
Canada
ContactUsLinkComponent
HighSpec Rho Starter Set 1: HighSpec Rho1D4 Agarose + Rho1D4 Peptide
HighSpec Rho Starter Set 1: HighSpec Rho1D4 Agarose + Rho1D4 Peptide
Catalog # 77393-070
Supplier:  CUBE BIOTECH
CAS Number:  
undefined
HighSpec Rho Starter Set 1: HighSpec Rho1D4 Agarose + Rho1D4 Peptide
Catalog # 77393-070
Supplier:  CUBE BIOTECH
Supplier Number:  33199
CAS Number:  

Specifications

  • Size:
    1 ml + 5 mg
  • Cat. no.:
    77393-070

Specifications

About this item

HighSpec Rho1D4 agarose resin is a suspension of 40 µm agarose beads, coupled to an anti-Rho1D4 antibody. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen.

  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead ligand: anti-Rho1D4 antibody
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C

Our Rho1D4 starter set 1 is a combination of our PureCube Rho1D4 agarose resin and the Rho1D4 elution peptide. It is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen.