Order Entry
Canada
ContactUsLinkComponent
 
Corrosive
Irritant
Environment
Ammonia solution 20 - 22%, OmniTrace Ultra™ for trace metal analysis, Supelco®
Catalog # CAAX1308-7
Supplier:  MilliporeSigma
CAS Number:  
Corrosive
Irritant
Environment
Ammonia solution 20 - 22%, OmniTrace Ultra™ for trace metal analysis, Supelco®
Catalog # CAAX1308-7
Supplier:  MilliporeSigma
Supplier Number:  AX1308-7
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    PTFE bottle
  • Size:
    250 ml
  • Cat. no.:
    CAAX1308-7
  • Supplier No.:
    AX1308-7

Specifications

Specification Test Results

  • Aluminum (Al):
    20pg/g max
  • Antimony (Sb):
    10pg/g max
  • Arsenic (As):
    10pg/g max
  • Assay (as NH3):
    20-22%
  • Barium (Ba):
    10pg/g max
  • Beryllium (Be):
    10pg/g max
  • Bismuth (Bi):
    10pg/g max
  • Cadmium (Cd):
    10pg/g max
  • Calcium (Ca):
    10pg/g max
  • Cerium (Ce):
    10pg/g max
  • Cesium (Cs):
    10pg/g max
  • Chromium (Cr):
    10pg/g max
  • Cobalt (Co):
    10pg/g max
  • Copper (Cu):
    10pg/g max
  • Dysprosium (Dy):
    10pg/g max
  • Erbium (Er):
    10pg/g max
  • Europium (Eu):
    10pg/g max
  • Gadolinium (Gd):
    10pg/g max
  • Gallium (Ga):
    10pg/g max
  • Germanium (Ge):
    10pg/g max
  • Gold (Au):
    10pg/g max
  • Holmium (Ho):
    10pg/g max
  • Indium (In):
    10pg/g max
  • Iron (Fe):
    10pg/g max
  • Lanthanum (La):
    10pg/g max
  • Lead (Pb):
    10pg/g max
  • Lithium (Li):
    10pg/g max
  • Lutetium (Lu):
    10pg/g max
  • Magnesium (Mg):
    10pg/g max
  • Manganese (Mn):
    10pg/g max
  • Mercury (Hg):
    200pg/g max
  • Molybdenum (Mo):
    10pg/g max
  • Neodymium (Nd):
    10pg/g max
  • Nickel (Ni):
    10pg/g max
  • Niobium (Nb):
    10pg/g max
  • Potassium (K):
    10pg/g max
  • Praseodymium (Pr):
    10pg/g max
  • Rhodium (Rh):
    10pg/g max
  • Rubidium (Rb):
    10pg/g max
  • Samarium (Sm):
    10pg/g max
  • Scandium (Sc):
    10pg/g max
  • Silver (Ag):
    10pg/g max
  • Sodium (N/A):
    20pg/g max
  • Strontium (Sr):
    10pg/g max
  • Tellurium (Te):
    10pg/g max
  • Terbium (Tb):
    10pg/g max
  • Thallium (Tl):
    10pg/g max
  • Thorium (Th):
    10pg/g max
  • Thulium (Tm):
    10pg/g max
  • Tin (Sn):
    10pg/g max
  • Titanium (Ti):
    10pg/g max
  • Tungsten (W):
    10pg/g max
  • Uranium (U):
    10pg/g max
  • VaN/Adium (V):
    10pg/g max
  • Ytterbium (Yb):
    10pg/g max
  • Yttrium (Y):
    10pg/g max
  • Zinc (Zn):
    10pg/g max
  • Zirconium (Zr):
    10pg/g max

Specification Test Results

About this item

 :
 :35.05 g/mol
Density:0.92 g/cm³ (20 °C)
UN:2672
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:215-647-6
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
ADR:8,