

Acetone ≥99.5% (by GC, corrected for water content) for pesticide residue analysis, Supelco®
Catalog # CAAX0114-1
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 solvent selection guide recommended after discussion
- Cat. no.:CAAX0114-1
- Supplier No.:AX0114-1
Specifications
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765