Order Entry
Canada
ContactUsLinkComponent
Bottle Brush, Rounded Tip, Justman Brush
Bottle Brush, Rounded Tip, Justman Brush
Catalog # 17140-004
Supplier:  Justman Brush
undefined
Bottle Brush, Rounded Tip, Justman Brush
Catalog # 17140-004
Supplier:  Justman Brush
Supplier Number:  J720373

Specifications

  • Bristle Material:
    Natural fiber
  • Handle Material:
    Wire
  • Total length:
    27.9 cm (11")
  • Brush Head Width:
    3.5 cm (1 3/8 ")
  • Cat. no.:
    17140-004
  • Supplier No.:
    5208
  • Brush Head Length:
    9.2 cm (3 5/8")

Specifications

About this item

Natural black bristles and rounded tip.

Safety ridges on the steel handle provide a good grip when washing glassware.