Orders LinkContactUsLinkComponent
VITLAB® Forceps, PMP, BrandTech
 :   Brandtech
VITLAB® Forceps, PMP, BrandTech
 :  Brandtech
VITLAB® Forceps, PMP, BrandTech
VITLAB® Forceps, PMP, BrandTech
Polymethylpentene (PMP) forceps with pointed ends.
  • Autoclavable at 121 °C for 20 minutes
  • Translucent
PMP is suitable for autoclaving and microwaves. Resistant to aliphatic alcohols, aldehydes, ester, aliphatic hydrocarbons, ketones, alkalis, and most acids (except oxidizing acids). Not recommended for use with ether, aromatic hydrocarbons, halogenated hydrocarbons, and oxidizing agents. Color: transparent white.
Operating temperature: 0 to 150°C (32 to 302°F)
Product Family Options
  • Length
  • 115 mm (4½")
    Pack of 10
    Pack of 10 - $115.79
    $115.79
    Pack of 10
       $115.79
     : 67895
  • 145 mm (5¾")
    Pack of 10
    Pack of 10 - $132.95
    $132.95
    Pack of 10
       $132.95
     : 67995