Reagent Reservoir, 5 ml, Sterile
Catalog #: 77395-248
Supplier: Heathrow Scientific
New Product
Specifications
- Sterility:Sterile
- Material:PS
- Cat. no.:77395-248
- Packaging:Single wrap
Specifications
About this item
Reagent Reservoirs by Heathrow Scientific offer a fresh and modern design with enhanced features to streamline pipetting procedures.
- Superior durability with enhanced walls and crush-resistant 'turrets' at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
- Graduation markings on the top edge and inner walls allow measurements of liquid to be easily identified and reduce reagent waste
- Pour-off spouts conveniently located on all four corners for easy spill-free pouring and reduced dripping
- Divided reservoir design allows pipetting of two different reagents; maximize recovery if using only single channel and small amount of reagent
Sterile reagent reservoirs are manufactured and gamma irradiated in an ISO certified facility. They are certified Dnase-, Rnase-, and pyrogen-free.
Packaged individually in an easy open HDPE package. Traceability Certificate available upon request.
Customers Who Bought This Also Bought