Vortex Genie® MAX Mixers, 230 V
Supplier: Scientific Industries
Certificates
About this item
Vortex Genie® MAX mixers are great for heavy duty high velocity vortex mixing.
- 230 V model
- High velocity
- Heavy zinc die casting
- Prevents 'walking'
- Ratcheting clamping system
- Ideal for repetitive protocols and use with open containers
Mixers are fully programmable for speed and time, up to 2000 rpm. They feature durable and heavy zinc die casting with specially designed feet prevents 'walking'. Holds six 250 ml flasks (no additional holder necessary), tube racks can hold up to 20 tubes, and two 2-MicroPlate inserts holding up to 4 microplates. Unique ratcheting clamping system will securely hold flasks, bottles, tubes and microplates.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (10790-358) is also available, please see separate product page.
Vortex Genie® MAX Components and Accessories
Product Information
Frequently Bought Together