

Acetone ≥99.9% (dried basis), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Catalog # CABJGC010-4
Supplier: Burdick & Jackson
CAS Number:
Specifications
- Pack type:Glass bottle, amber
- Size:4 L
- Cat. no.:CABJGC010-4
- Supplier No.:GC010-4
Specifications
Specification Test Results
- Electron Capture GC (as lindane):2 ppt max.
- Flame Ionization GC (as nonane):1 ppb max.
- GC Analysis (excluding water):99.9 % min.
- Refractive Index (20ºC):1.3583 - 1.3589
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.5 % max.
Specification Test Results
About this item
:
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56 °C
Melting Pt:–94.8 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS07
:GHS02
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765