Digital Mini Rotators, 230 V
Catalog # 76417-542
Supplier: Thermo Fisher Scientific
Irregular Voltage
Specifications
- Description:Digital mini rotator, EU/UK/CHN plug
- Platform dimensions:310×284 mm (12.2×11.2")
- Control:LED
- Display:LED
- Speed accuracy:±1 rpm (≤100rpm); ±1% (101 - 500rpm)
- Platform material:Aluminum
- Cat. no.:76417-542
- L×W×H:487×387×264 mm (19.2×15.2×10.4")
- Electrical:230 V, 50/60 Hz, 40 VA
- Humidity Range:20 - 85% RH
- Speed Range:50 - 500 rpm
- Maximum Load:4.5 kg
- Orbit:Ø 10 mm (0.4")
- Timer:1 min - 99 h 59 min or continuous
- Weight:4.5 kg (9.92 lbs.)
Specifications
About this item
The Thermo Scientific™ Digital mini rotator has been designed for culturing bacteria and fungi, hybridization assays, and gel staining and de-staining.
- Continuous operation or timer modes
Provides smooth and accurate orbital mixing of various vessels in the laboratory.