Digital Rockers, 230 V
Catalog #: 76417-530
Supplier: Thermo Fisher Scientific
Irregular Voltage
Specifications
- Description:Digital rocker, EU/UK/CHN Plug
- Max. load:4.5 kg (9.92 lbs.)
- Control:LED
- Display:LED
- Speed accuracy:±1 rpm ≤50 rpm; ±2 rpm >50rpm
- Timer:0 (continuous)/1 s - 9999 min.
- Weight:13.5 kg (29.8 lbs.)
- Platform material:Stainless steel
- Cat. no.:76417-530
- L×W×H:487×387×354 mm (19.2×15.2×13.9")
- Electrical:230 V, 50/60 Hz, 0.1 A
- Humidity Range:20 - 85% RH
- Speed Range:1 - 70 rpm
- Rotation Angle:1 - 15°
- Platform Dimensions:322×282×19 mm (12.7×11.1×0.7")
Specifications
About this item
Digital Rocker is designed for ELISA, protein synthesis, gel staining and de-staining, antibody staining, and other time-sensitive applications.
- Digital display of speed and time
- Platform style: Tray
- Capacity: 6×96-well plates
- Timer range: Continuous or 1 s to 9999 minutes
Digital display with big platform, accurate speed control and quick angle adjustment for easy sample agitation. Optional non-slip rubber mat tray is designed for carrying plates, dishes, culture bottles and bags.
Certifications: cCSAus, CE, RoHS.
Ordering information: Two year warranty.
Delivery: Includes dimpled rubber mat tray, rubber strip, user manual, EU plug, UK plug, CHN plug, US/Japan plug.
Customers Who Bought This Also Bought