Order Entry
Canada
ContactUsLinkComponent
Shaker with 180° Shaking Motion, 240 V
Shaker with 180° Shaking Motion, 240 V
Catalog # 33725-048
Supplier:  Glas-Col
Irregular Voltage
Small Business Enterprise
undefined
Shaker with 180° Shaking Motion, 240 V
Catalog # 33725-048
Supplier:  Glas-Col
Supplier Number:  099AWS18024
Irregular Voltage
Small Business Enterprise

Specifications

  • Shaking motion:
    180 degree shaking motion
  • Shipping weight:
    11 lbs.
  • Speed:
    2-30 rpm
  • Cat. no.:
    33725-048
  • Electrical:
    240 V
  • Supplier No.:
    099AWS18024

Specifications

About this item

Simulates hand shaking by rotating through 180° arc.

  • 240 V model
  • Shakes two flasks simutaneously
  • Bench top mounting
  • Use with screw cap or side arm flasks

For use with small to medium peptide synthesis flasks and culture flasks (up to 2-¹/₄" diameter, 10" long). This shaker offers 180 degree agitation shaking for solid phase peptide synthesis and for hard to dissolve solids such as polymers.

Ordering information: Shaker base with 2 large finger clamps.
Delivery: 7 to 10 days after order.
Caution: 240 V model. Corresponding model with voltage suitable for use in US (33725-050) is also available, please see separate product page.