Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Variable Speed Rocker, 230 V

VWR® Variable Speed Rocker, 230 V

2D rocker, 230 V, 35.6×30.5 mm (14×12")
Catalog # 76471-706
Irregular Voltage
Supplier #:  BR2000-GME
VWR® Variable Speed Rocker, 230 V
Catalog # 76471-706
Irregular Voltage
VWR® Variable Speed Rocker, 230 V
VWR® Variable Speed Rocker, 230 V

2D rocker, 230 V, 35.6×30.5 mm (14×12")

Price
$1,228.09
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Designed to provide precise speed and tilt angles for a broad range of molecular and biological mixing applications.

  • 230 V model
  • Variable speed control up to 32 rpm
  • Easily adjustable tilt angle, from 0 to 30°
  • Large 14×12" work surface incudes non-skid rubber mat
  • Suitable for use in a cold room
  • Optional stacking tray
  • Two year warranty

With variable speed control and adjustable tilt angles, the VWR® Rocker is ideal for many different applications including, gel staining, washing, antibody staining and hybridizations.

A large platform with a non-skid surface is included, ideal for securing all popular laboratory vessels. A powerful brushless motor drive provides smooth and consistent rocking motion and speed is adjustable to provide rocking from gentle washing to aggressive mixing.

Certifications: CE Marked.

Ordering information: Optional 14×12" stacking platform is available. VWR® 2D Variable Speed Rocker comes with a two-year warranty.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (75832-308) is also available, please see separate product page.

Specifications
  • Description:
    2D rocker
  • Platform Size:
    35.6×30.5 mm (14×12")
  • Voltage:
    230 V
  • Cat. No.:
    76471-706
  • Tilt angle:
    Variable: 0 - ±30°
  • Speed:
    Variable 2 - 30 rpm
  • Load capacity:
    3.6 kg (8 lb.)
  • Dimensions:
    14×12×8"
  • Electrical:
    230 V
  • Weight:
    7.5 lbs.
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Optional Stacking Platform with Flat Mat, 3" Stacking Clearance, 12 x 12 x 3"
    Optional Stacking Platform with Flat Mat, 3" Stacking Clearance, 12 x 12 x 3"
    75832-310
    Log in for availability
    Each - $353.71

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.