ContactUsLinkComponent
Hanna Low Range Fluoride Colorimeter
Hanna Low Range Fluoride Colorimeter
  CA470355-122
 :  Hanna
New Product
undefined
Hanna Low Range Fluoride Colorimeter
  CA470355-122
 :  Hanna
 :  HI729
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Low range fluoride colorimeter
  • Environmentally Preferable:
  • Cat. no.:
    CA470355-122
Provides a simple, accurate and cost-effective way to measure fluoride.
  • Our portable handheld colorimeter goes where you go
  • Get simple results with easy one-button operation
  • Don't waste time matching to a color chart anymore, get direct results quickly
Improve the accuracy of your water testing results with our simple handheld colorimeter. With our low range fluoride colorimeter you achieve more precise technology than your standard chemical test kit. You won't have to struggle to determine colors visually, our handheld colorimeter eliminates the hassle of matching to a color chart by providing a direct digital fluoride result.