Specifications
- Volume:2 - 1000 μl
- Description:Pipette starter kit
- Cat. no.:77395-240
- No. of Channels:1
Specifications
About this item
Universal carousel pipette stand. The pipette starter kit is convenient for new labs or for customers who want a new set of pipettors covering the most commonly used volumes.
- Pearl® pipettes by Heathrow Scientific were specifically engineered for ease of use and comfort
- The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI)
- The pipettes require ²/₃ the operation pressure of most standard pipettes
- Each pipette calibrated and a calibration certificate is supplied
- Carousel pipette stand: Assorted colors purple, yellow, red, orange, green, grey, clear, and blue
- Pipettes: Purple, yellow, and blue
- Reservoirs: White
Hold and store pipettes in style with the Universal Carousel Pipette Stand by Heathrow Scientific that holds up to 8 single-channel, 4 multichannel, or a combination of both pipettes.
A sleek and modern design enables this stand to securely hold virtually every known brand of pipette in the marketplace making it truly ‘universal’ while adding a pop of color to a bench top Reagent Reservoirs.
Multi-channel reservoirs by Heathrow Scientific are superior in every way with thicker walls, crush-resistant 'turrets' at the corners, easy pour-spouts, 25 ml graduation marks, and tabs to prevent stacked reservoirs from sticking together.
Pipette stand: ABS
Pearl® pipette: ABS
Reagent reservoirs: PS