Order Entry
Canada
Orders LinkContactUsLinkComponent
Assay/Button Brush, Mill-Rose

Assay/Button Brush, Mill-Rose

Nylon, 16 mm (3/5"), 11.4 cm (5")
Catalog # 17210-008
Supplier:   Mill-Rose
Supplier #:  MR-8888
Assay/Button Brush, Mill-Rose
Catalog # 17210-008
Supplier:  Mill-Rose
Assay/Button Brush, Mill-Rose
Assay/Button Brush, Mill-Rose

Nylon, 16 mm (3/5"), 11.4 cm (5")

Price
$68.90
Log in for availability
Each Pack of 12
Product Details & Documents

Stiff black nylon bristles for quick cleaning button samples for assays.

Flat-trimmed brushes have 0.014 gauge bristles mounted within a stainless steel tube. Double brush head.

Specifications
  • Bristle material:
    Nylon
  • Brush head Ø:
    16 mm (3/5")
  • Total length:
    11.4 cm (5")