Heating Shaking Dry Bath Incubator, 230 V
Catalog #: 10054-218
Supplier: Thermo Fisher Scientific
Irregular Voltage
Specifications
- Temperature Range:Ambient +5 to 100C
- Description:Shaking Heating Dry Bath
- Electrical:230 V (multiple export plugs)
- Speed Range:50 - 1500 rpm (block dependent)
- Orbit:2 mm (0.08")
- Supplier No.:88880028
Specifications
About this item
Increase lab versatility with the Thermo Scientific™ Digital Heating Shaking Drybath, easy to use and can be used as a drybath, shaker or both.
- 230 V model
- PID control ensures consistent shaking results and precise temperature control to 0.1 degree
- Speed range from 150 to 1500 rpm (block dependent)
- Program Storage 10 programs, up to 10 steps per program
- Interval shaking function allows shaking and non-shaking in same program
- Two types of timing modes:
- Start only after temperature reaches desired set point
- Start immediately after the timer is set
- Low-profile design
- Easily changed aluminum blocks in multiple sizes allow for maximum flexibility in vessel choice
- Over temperature cut off safety protection
- Orbital diameter: (mm/Inch) 2/0.08
- Uniformity(±°C/°F) 0.5/0.9 at 80 °C
- Stability(±°C/°F) 0.15/0.27 at 80 °C
Precisely control both temperature and shaking speed digitally, for accurate repeatable results. Precise accuracy is ensured (0.1 °C) by its PID controller and timer. Usable with 96-well, 0.5 ml, 1.5 ml, and 50 ml tubes, blocks purchased separately. Wide Temperature range, Ambient +5 to –100 ℃ covers a broad range of applications including thawing, incubation, enzyme reactions, blood urea nitrogen determinations, and immunoassays.
Certifications: CE
Ordering information: Must purchase a block with unit.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (10054-216) is also available, please see separate product page.
Customers Who Bought This Also Bought