Orders LinkContactUsLinkComponent
ScrubberStand™ Stackable Stands
ScrubberStand™ Stackable Stands
Supplier:   Labconco
ScrubberStand™ Stackable Stands
Supplier:  Labconco
Certificates
ScrubberStand™ Stackable Stands
ScrubberStand™ Stackable Stands
2 Options Available Below
Product Details & Documents
Double your washing capacity by stacking 2 glassware washers, reduce floor space.
  • Space efficiency: Optimizes space by enabling vertical stacking of washers
  • Increased capacity: Doubles washing capacity without expanding laboratory area
  • Safe: ETL certification eliminates any worries over the possibility of tipping
  • Ease of installation: Comes already assembled
  • Compatibility: Designed to work seamlessly with our existing range of glassware washers
Maximize glassware washing capacity without using extra floor space. Stack 2 Freestanding Washers with a Stackable ScrubberStand™. Made of 1.5” tubular, powder-coated steel for durability and stability. Hardware included. Seismic bracket kit available.
Product Family Options
Product Information
  • For use with
    Availability
    Price
  • 77400-898
    Stackable ScrubberStand™
    Stackable ScrubberStand™
    For FlaskScrubber™ and SteamScrubber™ glassware washers
    Log in for availability
    Log in for availability
    Each
    Each - $1,540.00
    $1,540.00
    Each
       $1,540.00
    Minimum order quantity is 1
    Supplier Number: 4618000
  • 77400-900
    Stackable ScrubberStand™
    Stackable ScrubberStand™
    For FlaskScrubber™ Vantage glassware washers
    Log in for availability
    Log in for availability
    Each
    Each - $1,660.00
    $1,660.00
    Each
       $1,660.00
    Minimum order quantity is 1
    Supplier Number: 4618001
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.