Ammonia solution 20 - 22%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Catalog # CA87003-656
Supplier: BDH
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:HDPE bottle
- Size:1 L
- Cat. no.:CA87003-656
- Supplier No.:20701-VWNJ05
Specifications
Specification Test Results
- Assay:20 - 22%
- Aluminum (Al):max. 20 ppt
- Antimony (Sb):max. 10 ppt
- Arsenic (As):max. 10 ppt
- Barium (Ba):max. 10 ppt
- Beryllium (Be):max. 10 ppt
- Bismuth (Bi):max. 10 ppt
- Cadmium (Cd):max. 10 ppt
- Calcium (Ca):max. 10 ppt
- Cerium (Ce):max. 10 ppt
- Cesium (Cs):max. 10 ppt
- Chromium (Cr):max. 10 ppt
- Cobalt (Co):max. 10 ppt
- Copper (Cu):max. 10 ppt
- Dysprosium (Dy):max. 10 ppt
- Erbium (Er):max. 10 ppt
- Europium (Eu):max. 10 ppt
- Gadolinium (Gd):max. 10 ppt
- Gallium (Ga):max. 10 ppt
- Germanium (Ge):max. 10 ppt
- Gold (Au):max. 10 ppt
- Hafnium (Hf):Information Only
- Holmium (Ho):max. 10 ppt
- Indium (In):max. 10 ppt
- Iron (Fe):max. 10 ppt
- Lanthanum (La):max. 10 ppt
- Lead (Pb):max. 10 ppt
- Lithium (Li):max. 10 ppt
- Lutetium (Lu):max. 10 ppt
- Magnesium (Mg):max. 10 ppt
- Manganese (Mn):max. 10 ppt
- Mercury (Hg):max. 200 ppt
- Molybdenum (Mo):max. 10 ppt
- Neodymium (Nd):max. 10 ppt
- Nickel (Ni):max. 10 ppt
- Niobium (Nb):max. 10 ppt
- Palladium (Pd):Information Only
- Platinum (Pt):Information Only
- Potassium (K):max. 10 ppt
- Praseodymium (Pr):max. 10 ppt
- Rhenium (Re):Information Only
- Rhodium (Rh):max. 10 ppt
- Rubidium (Rb):max. 10 ppt
- Ruthenium (Ru):Information Only
- Samarium (Sm):max. 10 ppt
- Scandium (Sc):max. 10 ppt
- Selenium (Se):Information Only
- Silver (Ag):max. 10 ppt
- Sodium (Na):max. 20 ppt
- Strontium (Sr):max. 10 ppt
- Tellurium (Te):max. 10 ppt
- Terbium (Tb):max. 10 ppt
- Thallium (Tl):max. 10 ppt
- Thorium (Th):max. 10 ppt
- Thulium (Tm):max. 10 ppt
- Tin (Sn):max. 10 ppt
- Titanium (Ti):max. 10 ppt
- Tungsten (W):max. 10 ppt
- Uranium (U):max. 10 ppt
- Vanadium (V):max. 10 ppt
- Ytterbium (Yb):max. 10 ppt
- Yttrium (Y):max. 10 ppt
- Zinc (Zn):max. 10 ppt
- Zirconium (Zr):max. 10 ppt
Specification Test Results
About this item
:
:35.05 g/mol
Density:0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650