Order Entry
Canada
ContactUsLinkComponent
Acetone ≥99.5% Dehydrant, Epredia™ HiPur™ for histology
Flammable
Irritant
Acetone ≥99.5% Dehydrant, Epredia™ HiPur™ for histology
Catalog # CA48218-692
Supplier:  Thermo Scientific
CAS Number:  
undefined
Flammable
Irritant
Acetone ≥99.5% Dehydrant, Epredia™ HiPur™ for histology
Catalog # CA48218-692
Supplier:  Thermo Scientific
Supplier Number:  9000-1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 gal.
  • Cat. no.:
    CA48218-692
  • Supplier No.:
    9000-1

Specifications

Specification Test Results

  • Assay:
    99.5% min.

Specification Test Results

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
 :GHS02
 :GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765