SP Bel-Art Sterileware® Samplit™ Scoop and Container System, Polystyrene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
This collection device incorporates a sturdy sampling scoop that threads into a dedicated leakproof 190 mL (6.5oz.) container ensuring safe and efficient isolation of material.
- Sturdy polystyrene construction
- Dedicated leakproof container ensures efficient isolation
- Rigid scoop strong enough for semi-solids and granular materials
- Ribbed handle for excellent grip
The rigid scoop is made of white, high-impact polystyrene plastic which is strong enough for coring semi-solids such as cheese, ice cream, and even soil or granular materials.
A ribbed handle provides an excellent grip for pushing and turning the scoop. The depth and shape of the scoop keeps samples contained while allowing for easy dispensing into the clear styrene container. Supplied with labels for sample identification.
Rack 60916-260 can be used to store and transport Samplit™ containers.
Ordering information: Available in a sterile pack of 25, a nonsterile pack of 25, or a nonsterile scoops only pack of 25.