Wrist Action Shakers, 220 V
Supplier: Burrell Scientific
Certificates
About this item
Wrist action shaker, with 385 oscillations per minute (+/-) 10%.
- 220 V model
Max. load tested at 10 lbs. blue/white color.
Certifications: Complies with UL, CSA and CE.
Ordering information: Each wrist shaker comes with enough clamps to accommodate its full capacity of vessels. Optional top platform accessory accommodates up to eight 250 ml flasks and attaches directly to main shaft of shaker.
Caution: 220 V model. Corresponding models with voltage suitable for use in US (10161-186, 57040-027, 57040-049, 57040-060, 57040-082) are also available, please see separate product page.
Customers Who Bought This Also Bought