Standard MA-P Precision Balances, Legal for Trade
Supplier: Mettler Toledo
New Product
Certificates
About this item
METTLER TOLEDO Standard MA-P Legal for Trade Precision Balances are simple to operate and portable for on-the-go weighing, without compromising on accuracy or performance.
- Strain gauge weighing cell
- Internal adjustment
- LCD hybrid touchscreen
- 10 built-in applications
- Power cable and battery operated
- Front level indicator; overload protection
- 2 interfaces: USB-A, RS232
- Bluetooth option
The precise weigh cell delivers highly accurate results with each use. Increase efficiency in the workflow with the intuitive touchscreen that guides you through each weighing procedure. Easily export data to a connected printer, computer, or USB device.
Certifications: OIML/NTEP approved.
Specifications
- Catalog No:
- 77323-262
- 77323-266
- 77323-270
- Communication interface:
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- Voltage:
- 100 - 240 V, 50/60 Hz or 8 AA batteries
- 100 - 240 V, 50/60 Hz or 8 AA batteries
- 100 - 240 V, 50/60 Hz or 8 AA batteries