Standard MA Analytical Balances
Supplier: Mettler Toledo
New Product
Certificates
About this item
METTLER TOLEDO Standard MA Analytical Balances are highly accurate, efficient, simple to operate and designed with robust construction for proven long lasting performance.
- Internal adjustment
- Front level indicator
- Metal base, PBT top housing
- Overload protection
- 2 interfaces: USB-A, RS232
- Bluetooth option
The LCD hybrid touchscreen is simple to operate and will increase efficiency of daily tasks with 9 built-in applications and statistical data analysis. Built with a metal base, PBT top housing and overload protection to defend against rough handling in any weighing environment, ensuring quality performance for years to come.
Specifications
- Catalog No:
- 77323-192
- 77323-272
- 77323-196
- CA77323-188
- CA77323-192
- CA77323-196
- CA77323-180
- CA77323-184
- 77323-180
- 77323-184
- CA77323-272
- 77323-188
- Communication interface:
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz