Orders LinkContactUsLinkComponent
VWR® Dumont Forceps
VWR® Dumont Forceps
114 mm (4½")
Catalog # 82027-426
Supplier:   VWR International
VWR® Dumont Forceps
Catalog # 82027-426
Supplier:  VWR International
VWR® Dumont Forceps
VWR® Dumont Forceps
114 mm (4½")
Price
$50.01
New Customer Promotional Price
Log in for availability
Each Pack of 12 Case of 60
Product Details & Documents
Ideal for use with specimen screens for electron microscopes and electronic parts.
  • Stainless steel fabrication
  • Sharpened to a very fine point
Dumont forceps are ergonomically designed for comfortable use in a variety of applications.
Documents
Specifications
  • Length:
    114 mm (4½")