ContactUsLinkComponent
Glass Cutters, Tungsten Carbide
Glass Cutters, Tungsten Carbide
Catalog # 32876-021
Supplier:  Tool Specialty
undefined
Glass Cutters, Tungsten Carbide
Catalog # 32876-021
Supplier:  Tool Specialty
Supplier Number:  SK-05180
Specifications
  • Description:
    Glass Cutters
  • Material:
    Tungsten carbide
  • Cat. no.:
    32876-021
  • Supplier No.:
    SK-05180
  • Grip Type:
    Wood
Specifications
About this item
The speciality glass cutters used to cutting glass tubing.
Designed with steel blade with cutting edges tipped with tungsten carbide.