Chemware® Micro Spatula, Teflon® Resin-Coated, Saint-Gobain Performance Plastics
Catalog # 57949-033
Supplier: Saint Gobain Performance Plastics
Specifications
- Length:15.2 cm (6")
- Cat. no.:57949-033
- Supplier No.:D1069292
Specifications
About this item
Both ends have a durable Teflon® resin coating, which gives an inert surface for handling chemicals and prevents loss due to sticking.
- Hayman style