ContactUsLinkComponent
SP Bel-Art Stainless Steel Reagent Digger, Bel-Art Products, a part of SP
  62489-110
undefined
SP Bel-Art Stainless Steel Reagent Digger, Bel-Art Products, a part of SP
  62489-110
 :  F36713-0000
  • Length:
    22.9L cm (9")
  • Cat. no.:
    62489-110
  • Supplier No.:
    367130000
Ideal for dislodging and retrieving stubborn, caked, or crystallized materials from deep down inside a bottle or vessel that has a minimum neck diameter of 24 mm (15/16").
  • Ergonomically designed
  • Suitable for a variety of reagent applications
  • Stainless steel construction
  • Ideal for removing stubborn materials from deep vessels
This utensil is ergonomically designed with a curved shape, one pointed end, and one blunt end. The shape facilitates scooping, shoveling, digging, and transferring of powdered, flaked, crystalline, or granular media.