ContactUsLinkComponent
D-PASMC – Diseased Human Pulmonary Artery Smooth Muscle Cells – Diabetes Type II
D-PASMC – Diseased Human Pulmonary Artery Smooth Muscle Cells – Diabetes Type II
Catalog # CA10064-306
CAS Number:  
undefined
D-PASMC – Diseased Human Pulmonary Artery Smooth Muscle Cells – Diabetes Type II
Catalog # CA10064-306
Supplier Number:  CC-2913
CAS Number:  
Specifications
  • Description:
    D-PASMC – Diseased Human Pulmonary Artery Smooth Muscle Cells – diabetes type II
  • Size:
    ≥500000 cells
  • Cat. no.:
    CA10064-306
Specifications
About this item
Pulmonary artery smooth muscle cells are isolated from human donors that have been diagnosed with diabetes type I or type II disease.
Diseased cells are grown in the same optimized media system as normal human pulmonary artery smooth muscle cells. Diabetic PASMC cultures can be used for experimental applications in diabetic-related cardiovascular pharmaceutical development, vascular pathology, including atherosclerosis.
Clonetics® D-PASMC diseased human pulmonary artery smooth muscle cells and media are convenient and easy to use, allowing the researcher to focus on results. Cryopreserved Clonetics® PASMC are shipped in third passage. Proliferating Clonetics® PASMC are shipped in fourth passage. Clonetics® cells, medium and reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.
Delivery: Content: 1×cryopreserved ampule with ≥500000 cells/vial.