Order Entry
Canada
ContactUsLinkComponent
Nucleofector® P5 Primary Cell 4D-X Kit S
Nucleofector® P5 Primary Cell 4D-X Kit S
Catalog # CA10064-526
CAS Number:  
Nucleofector® P5 Primary Cell 4D-X Kit S
Catalog # CA10064-526
Supplier Number:  V4XP-5032
CAS Number:  

Specifications

  • Description:
    P5 Primary Cell 4D-Nucleofector® X kit S
  • Environmentally Preferable:
  • Cat. no.:
    CA10064-526
  • No. of Reactions:
    32
  • Supplier No.:
    V4XP-5032

Specifications

About this item

The P5 primary cell 4D-Nucleofector® X kit S is one of our five 4D-Nucleofector® Kits suited for transfecting primary cells in the 20 µl 16-well Nucleocuvette® strip format when working with the 4D-Nucleofector® X unit.

  • High transfection efficiencies for primary cells
  • Cells maintain functionality post transfection
  • Conditions are transferable between 4D-Nucleofector® System, 96-well Shuttle® System and 384-well Nucleofector® System

This kit has been determined to be optimal for transfecting following cell types:
HUVEC
Skeletal Myoblast, hum.

Applications:
Transfection of low cell numbers in the range of 5×10⁴ to 1×10⁶ cells.
Medium throughput applications.
Optimization of transfection conditions e.g. substrate titrations.

Delivery: Content:
1×0.675 ml P5 primary cell Nucleofector® solution.
1×0.15 ml Supplement 3.
50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0).
2×16-well Nucleocuvette® strips (20 μl).