Septihol® Hand Sanitizer
Catalog #: 76499-224
Supplier: Steris
Specifications
- Capacity:266 ml (9 oz.)
- Description:Septihol® hand sanitizer
- Cat. no.:76499-224
- Packaging:Aerosol can
Specifications
About this item
Septihol® Hand Sanitizer is an alcohol-based hand rub that offers fast antimicrobial action on hands without the need for a water rinse.
- Fast acting, proven antimicrobial efficacy for hands
- Formulated with 62% (v/v) ethyl alcohol
- Lightweight foam mousse format allows the best possible hand coverage
- No rinse format with minimal residue on hands
- Manufactured under a cGMP, FDA compliant quality system
- Fragrance-free
Septihol® Hand Sanitizer is a luxurious foam sanitizer formulated with 62% v/v ethyl alcohol to provide a convenient method for routine hand washing. It is ideal for personnel in pharmaceutical environments to reduce the risk of environmental contamination. Septihol® Hand Sanitizer is an excellent adjunct to hand washing when additional antimicrobial protection is needed.
Packaging: 24 cans per case.
Customers Who Bought This Also Bought