ContactUsLinkComponent
Stilla® Ruby Chip
Stilla® Ruby Chip
  77392-266
 :  
undefined
Stilla® Ruby Chip
  77392-266
 :  C16011
 :  
  • Description:
    Ruby chip
  • Cat. no.:
    77392-266
  • Units:
    Pack of 12 chips
  • Details:
    Storage temperature: −20... ±5 °C
The Ruby Chip features cutting-edge, patented microfluidic technology that integrates the entire digital PCR workflow into a single fully automation-compatible consumable.
  • Capacity: 16 samples/chip
  • Input volume: 5 µl/chamber
  • Droplets per sample: Up to 17000
  • Dynamic range: ∼4.7 log
  • Fully compatible with integration to automated liquid handling equipment
For higher throughput applications, the Ruby Chip offers the analysis of up to 48 samples per run for your digital PCR workflows. After sample preparation, just a single pipetting step is required before the chip is loaded onto either the naica® system or the Nio™+ for partitioning, PCR amplification, and sample read-out. The Ruby Chip is also fully compatible with integration to automated liquid handling equipment.
Digital PCR Consumables and Reagents