ContactUsLinkComponent
SingleQuots® PGM-2 Preadipocyte Growth Medium- Supplements and Growth Factors
SingleQuots® PGM-2 Preadipocyte Growth Medium- Supplements and Growth Factors
Catalog # CA10064-102
CAS Number:  
undefined
SingleQuots® PGM-2 Preadipocyte Growth Medium- Supplements and Growth Factors
Catalog # CA10064-102
Supplier Number:  PT-9502
CAS Number:  
Specifications
  • Description:
    PGM-2 Preadipocyte growth medium Singlequots®- supplements and growth factors
  • Cat. no.:
    CA10064-102
  • Supplier No.:
    PT-9502
Specifications
About this item
PGM® -2 Preadipocyte Growth Medium-2 BulletKit® Medium provides both a proliferation medium and a differentiation medium guaranteed to induce differentiation into mature, functionally active adipocytes; Each bottle of the PGM-2® BulletKit® Medium contains enough differentiation medium to fully differentiate 10 million preadipocytes [10 complete 96-well plates] into mature, functionally active adipocytes.
The PGM®-2 Medium is offered as a BulletKit® Medium (catalog no. PT-8002) which includes both the basal medium and the necessary supplements for both proliferation and differentiation of preadipocytes. The basal medium (PGM®-2 Basal Medium; catalog no. PT-8202) and the necessary supplements (PGM®-2 SingleQuot® Kit; catalog no. PT-9502) can also be purchased individually.