ContactUsLinkComponent
BulletKit® PGM-2® Preadipocyte Growth Medium-2
BulletKit® PGM-2® Preadipocyte Growth Medium-2
Catalog # CA10064-402
CAS Number:  
undefined
BulletKit® PGM-2® Preadipocyte Growth Medium-2
Catalog # CA10064-402
Supplier Number:  PT-8002
CAS Number:  
Specifications
  • Description:
    PGM-2® Preadipocyte growth medium-2
  • Cat. no.:
    CA10064-402
Specifications
About this item
PGM® -2 Preadipocyte Growth Medium-2 BulletKit® provides both a proliferation medium and a differentiation medium guaranteed to induce differentiation into mature, functionally active adipocytes.
Each bottle of the PGM-2® BulletKit® Medium contains enough differentiation medium to fully differentiate 10 million preadipocytes [10 complete 96-well plates] into mature, functionally active adipocytes.
The PGM®-2 Medium is offered as a BulletKit® Medium (catalog no. PT-8002) which includes both the basal medium and the necessary supplements for both proliferation and differentiation of preadipocytes.
The basal medium (PGM®-2 Basal Medium; catalog no. PT-8202) and the necessary supplements (PGM®-2 SingleQuot® Kit; catalog no. PT-9502) can also be purchased individually.
Delivery: Content:
1 × PGM®-2 Basal Medium (PT-8202)
1 × PGM®-2 SingleQuots® Supplement and Growth Factor Pack (PT-9502)