ContactUsLinkComponent
SingleQuots® MSCGM® Supplements and Growth Factors
SingleQuots® MSCGM® Supplements and Growth Factors
Catalog # CA10063-958
CAS Number:  
undefined
SingleQuots® MSCGM® Supplements and Growth Factors
Catalog # CA10063-958
Supplier Number:  PT-4105
CAS Number:  
Specifications
  • Description:
    MSCGM® Singlequots® supplements and growth factors
  • Cat. no.:
    CA10063-958
  • Supplier No.:
    PT-4105
Specifications
About this item
The MSCGM hMSC SingleQuot Kit is a component of Lonza's MSCGM BulletKit (catalog no. PT-3001),a serum containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state.
The serum used in Lonza’s MSCGM®human Mesenchymal Stem Cell Growth Medium BulletKit® Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic, chondrogenic, or adipogenic lineages (when properly stimulated with an appropriate differentiation media).
Lonza guarantees the performance of Clonetics®/Poietics® cells only if appropriate Clonetics®/Poietics® media and reagents are used exclusively and the recommended storage and use protocols are followed.