ContactUsLinkComponent
ReagentPack® Subculture Reagents
ReagentPack® Subculture Reagents
Catalog #: CA71005-538
CAS Number:  
REAGENT PK SUBCULTURE REAGENTS
ReagentPack® Subculture Reagents
Catalog #: CA71005-538
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Reagentpack® Subculture reagents
  • Cat. no.:
    CA71005-538
  • Supplier No.:
    CC-5034
Specifications
About this item
ReagentPack® Subculture Kits have been designed specifically for the passaging of primary cell types. Each kit contains 100 mL each of: Trypsin/EDTA, Trypsin Neutralizing Solution, HEPES Buffered Saline.
The ReagentPack® Subculture Reagents kit is recommended for use with Lonza's primary cells to ensure effective trypsinization and a successful cell passaging process.
Delivery: Content:
1 × Bottle Trypsin/EDTA 100 ml
1 × Bottle Trypsin Neutralizing Solution 100 ml
1 × Bottle HEPES Buffered Saline Solution 100 ml
Customers Who Bought This Also Bought