Trypsin Neutralizing Solution
Catalog # CA10064-044
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Trypsin neutralizing solution
- Cat. no.:CA10064-044
- Supplier No.:CC-5002
Specifications
About this item
Trypsin Neutralizing Solution (TNS) is used to effectively neutralize Trypsin/EDTA. Lonza's TNS is availableindividually or as a component in our ReagentPack® Subculture Reagents (CC-5034) which includes Trypsin and HEPES buffer.
Delivery information: Content: 1 × Bottle Trypsin Neutralizing Solution, 100 ml