- Description:MGM®- 4 Melanocyte growth medium-4 Singlequots® supplements and growth factors
- Cat. no.:CA10064-014
- Supplier No.:CC-4435
MGM® -4 SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the MGM® Melanocyte Cell Growth Medium BulletKit® (Catalog Number CC-3249). The full MGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3249.
The MGM®-4 Melanocyte Growth Medium-4 BulletKit®is recommended media for melanocyte optimal growth and survival in culture. This medium is offered in our traditional BulletKit® format, providing flexibility to add or omit components as research needs dictate.
ET-3 Growth Supplement, an additional growth factor required for adult melanocyte growth, is sold separately to supplement the MGM®-4 BulletKit® for a complete formulation specifically designed for growing adult melanocytes. ET-3 offers enhanced growth and plating advantages in addition to limiting other cell type contamination in culture.
1 × Natural Cap Vial with CaCl₂ 1.00 ml
1 × Amber Vial with PMA 0.50 ml
1 × Gray Cap Vial with hFGF-B 1.00 ml
1 × Lilac Cap Vial with Insulin 1.00 ml
1 × Orange Cap Vial with BPE 2.00 ml
1 × Natural Cap Vial with Hydrocortisone 0.50 ml
1 × Bottle FBS 2.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml"