Ascorbic Acid
Catalog # CA10064-008
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Ascorbic acid
- Cat. no.:CA10064-008
Specifications
About this item
Ascorbic Acid is an important component in various primary cell culture media kits. It is included in various BulletKits. Ascorbic Acid is also sold separately at a concentration of 70 mM.
Delivery: Content: 1 × Vial Containing 70 mM Ascorbic Acid 0.5 ml.