SingleQuots® SCGM® Stromal Cell Growth Medium Supplements and Growth Factors
CA10063-950
:
- Description:SCGM® Stromal cell growth medium Singlequots® supplements and growth factors
- Cat. no.:CA10063-950
SCGM® SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the SCGM® Stromal Cell Growth Medium BulletKit® (Catalog Number CC-3205).
The full SCGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3205. Stromal Cell Growth Medium was developed to support the maintenance and expansion of prostate stromal cells and certain aortic and dental fibroblasts. SCGM®medium contains 5% FBS.
Clonetics® Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.
: Content: 1 × Lilac Cap Vial with Insulin 0.50 ml, 1 × Gray Cap Vial with hFGF-B 0.50 ml, 1 × Red Cap Vial with GA-1000 0.50 ml, 1 × Bottle FBS 25.00 ml.