ContactUsLinkComponent
SingleQuots® PrEGM® Prostate Epithelial Cell Growth Medium Supplements and Growth Factors
SingleQuots® PrEGM® Prostate Epithelial Cell Growth Medium Supplements and Growth Factors
Catalog # CA10063-948
CAS Number:  
undefined
SingleQuots® PrEGM® Prostate Epithelial Cell Growth Medium Supplements and Growth Factors
Catalog # CA10063-948
Supplier Number:  CC-4177
CAS Number:  
Specifications
  • Description:
    PrEGM® Prostate epithelial cell growth medium Singlequots® supplements and growth factors
  • Cat. no.:
    CA10063-948
Specifications
About this item
PrEGM® SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the PrEGM® Prostate Epithelial Cell Growth Medium BulletKit® (Catalog Number CC-3166).
The full PrEGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3166.
PrEGM® Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. PrEGM® Medium contains no serum.
Delivery: Content:
1 × Orange Cap Vial with BPE 2.00 ml
1 × Amber Vial with Triiodothyronine 0.50 ml
1 × Lilac Cap Vial with Insulin 0.50 ml
1 × Green Cap Vial with hEGF 0.50 ml
1 × Natural Cap Vial with Hydrocortisone 0.50 ml
1 × Natural Cap Vial with Transferrin 0.50 ml
1 × Amber Vial with Epinephrine 0.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml
1 × Amber Vial with Retinoic Acid 0.50 ml