SingleQuots® FGM®-2 Fibroblast Growth Medium-2 Supplements and Growth Factors
Catalog # CA10064-368
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:FGM®-2 Fibroblast growth medium-2 Singlequots® supplements and growth factors
- Cat. no.:CA10064-368
Specifications
About this item
FGM® -2 SingleQuot® Kit are components sold separately and does not include the basal medium. These are part of the FGM® -2 Fibroblast Growth Medium BulletKit® (Catalog Number CC-3132) which consists of basal medium and supplements packaged together.
FGM®-2 Fibroblast Growth Medium-2 BulletKit® was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM®-2 Medium contains 2% serum.
Delivery: Content:
1 × Lilac Cap Vial with Insulin 0.50 ml
1 × Gray Cap Vial with hFGF-B 0.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml
1 × Bottle FBS 10.00 ml
1 × Lilac Cap Vial with Insulin 0.50 ml
1 × Gray Cap Vial with hFGF-B 0.50 ml
1 × Red Cap Vial with GA-1000 0.50 ml
1 × Bottle FBS 10.00 ml