SingleQuots® SAGM® Small Airway Epithelial Cell Growth Medium Supplements and Growth Factors
Catalog # CA10064-366
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:SAGM® Small airway epithelial cell growth medium Singlequots® supplements and growth factors
- Cat. no.:CA10064-366
- Supplier No.:CC-4124
Specifications
About this item
SAGM® SingleQuot® Kits are components sold separately and does not include the basal medium. These are part of the full SAGM® Small Airway Epithelial Cell Growth Medium BulletKit® (catalog number CC-3118) which consists of basal medium and supplements packaged together. SAGM® BulletKit®.
SAGM® BulletKit® is serum-free and has been optimized to grow human small airway epithelial cells (SAEC).
Delivery: Content:
1 × Orange Cap Vial with BPE 2.0 ml
1 × Lilac Cap Vial with Insulin 0.5 ml
1 × Natural Cap Vial with Hydrocortisone 0.5 ml
1 × Red Cap Vial with GA-1000 0.5 ml
1 × Amber Vial with Retinoic Acid 0.5 ml
1 × Bottle with Fatty Acid Free BSA 5 ml
1 × Natural Cap Vial with Transferrin 0.5 ml
1 × Amber Vial with Triiodothyronine 0.5 ml
1 × Amber Vial with Epinephrine 0.5 ml
1 × Green Cap Vial with hEGF 0.5 ml
1 × Orange Cap Vial with BPE 2.0 ml
1 × Lilac Cap Vial with Insulin 0.5 ml
1 × Natural Cap Vial with Hydrocortisone 0.5 ml
1 × Red Cap Vial with GA-1000 0.5 ml
1 × Amber Vial with Retinoic Acid 0.5 ml
1 × Bottle with Fatty Acid Free BSA 5 ml
1 × Natural Cap Vial with Transferrin 0.5 ml
1 × Amber Vial with Triiodothyronine 0.5 ml
1 × Amber Vial with Epinephrine 0.5 ml
1 × Green Cap Vial with hEGF 0.5 ml