PNBM® Primary Neuron Basal Medium
Catalog # CA10063-918
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:PNBM® Primary neuron basal medium
- Cat. no.:CA10063-918
Specifications
About this item
Clonetics® PNBM® Primary Neuron Basal Medium (CC-3256) specifically developed for culturing embryonic primary rat and mouse neurons.For successful culture of neurons, basal media should be combined with growth factors (sold seperately).
- Serum-free
- Allows for long-term maintenance and growth of neuronal cells
- Promotes development of high quality neuronal networks without the need of an astrocyte feeder layer
Basal medium and singlequots growth factors can be purchased seperately or in convenient PNGM® BulletKit® format (CC-4461).
Delivery: Content: 1 × 200 ml Bottle Basal Medium (growth factors not included).