ReagentPack® Chondrocyte Subculture Reagents
Catalog # CA10063-896
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Chondrocyte Reagentpack® subculture reagents
- Cat. no.:CA10063-896
Specifications
About this item
Chondrocyte ReagentPack® Subculture Kits have been designed specifically for the passaging of primary chondrocytes.
The Chondrocyte ReagentPack® Subculture Kit is recommended for use with Lonza's primary chondrocytes for effective trypsinization and cell passaging process.
Delivery: Content:
1 × Bottle Trypsin/EDTA 100 ml
1 × Bottle Trypsin Neutralizing Solution 100 ml
1 × Bottle HEPES Buffered Saline Solution 100 ml
1 × Bottle Trypsin/EDTA 100 ml
1 × Bottle Trypsin Neutralizing Solution 100 ml
1 × Bottle HEPES Buffered Saline Solution 100 ml