CBM® Chondrocyte Growth Basal Medium
CA10064-360
:
- Cat. no.:CA10064-360
- Supplier No.:CC-3217
CBM® is a Chondrocyte cell growth basal medium sold separately. This can be used with 1 kit of CC-4409 (chondrocyte cell growth medium SingleQuots® Supplements and Growth Factors). The full CGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3216.
CGM® Chondrocyte Growth Medium is specially designed for expansion of chondrocytes. The BulletKit® is conveniently packaged with basal medium and supplements included for the growth of chondrocytes.
CDM® Chondrocyte Differentiation Medium is also available and sold separately for differentiation of human chondrocytes.
: Content: 1 × Bottle Basal Medium 500 ml.