BulletKit® NPMM® Neural Progenitor Maintenance Medium
CA10063-888
:
- Cat. no.:CA10063-888
Neural Progenitor Maintenance Medium (NPMM) BulletKit® contains the necessary supplements and media for optimal normal human neural progenitor cell growth and differentiation.
: Content:
1 × NPBM® Neural Progenitor Basal Medium (CC-3210), 200 ml
1 × NPMM® Neural Progenitor Maintenance Medium SingleQuots® Kit (CC-4241)
containing:
1 × hEGF, 0.40 ml
1 × hFGF, 0.40 ml
1 × NPMM® Neural Progenitor Supplement SingleQuots® Kit (CC-4242) containing:
1 × NSF-1, 4.0 ml
1 × GA-1000, 0.40 ml
1 × NPBM® Neural Progenitor Basal Medium (CC-3210), 200 ml
1 × NPMM® Neural Progenitor Maintenance Medium SingleQuots® Kit (CC-4241)
containing:
1 × hEGF, 0.40 ml
1 × hFGF, 0.40 ml
1 × NPMM® Neural Progenitor Supplement SingleQuots® Kit (CC-4242) containing:
1 × NSF-1, 4.0 ml
1 × GA-1000, 0.40 ml