BulletKit® SmGM®- 2 Smooth Muscle Cell Growth Medium -2
Catalog # CA71005-518
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:SmGM®- 2 Smooth muscle cell growth medium -2
- Cat. no.:CA71005-518
- Supplier No.:CC-3182
Specifications
About this item
SmGM®-2 Smooth Muscle Growth Medium-2 is a robust medium that was developed for the maintenance and expansion of human smooth muscle cells from various tissue origins including cardiac, airway, prostate and more.
This kit is also utilized to support the maintenance of intestinal cells. SmGM®-2 medium contains 5% FBS.
Clonetics® Media Products are specially designed to support the growth of human and animal primary derived cells. Media systems have been specifically developed and optimized for various cell systems.
Delivery: Content:
1 × SmBM® Basal Medium (CC-3181), 500 ml
1 × SmGM® -2 SingleQuots®Supplement Pack (CC-4149) containing:
1 × Lilac Cap Vial with Insulin, 0.50 ml
1 × Gray Cap Vial with hFGF-B, 1.00 ml
1 × Red Cap Vial with GA-1000, 0.50 ml
1 × Bottle FBS, 25.00 ml
1 × Green Cap Vial with hEGF, 0.50 ml
1 × SmBM® Basal Medium (CC-3181), 500 ml
1 × SmGM® -2 SingleQuots®Supplement Pack (CC-4149) containing:
1 × Lilac Cap Vial with Insulin, 0.50 ml
1 × Gray Cap Vial with hFGF-B, 1.00 ml
1 × Red Cap Vial with GA-1000, 0.50 ml
1 × Bottle FBS, 25.00 ml
1 × Green Cap Vial with hEGF, 0.50 ml