ContactUsLinkComponent
BEBM® Bronchial Epithelial Cell Growth Basal Medium
BEBM® Bronchial Epithelial Cell Growth Basal Medium
Catalog # CA11006-400
CAS Number:  
undefined
BEBM® Bronchial Epithelial Cell Growth Basal Medium
Catalog # CA11006-400
Supplier Number:  CC-3171
CAS Number:  
Specifications
  • Description:
    BEBM® bronchial epithelial cell growth basal medium
  • Cat. no.:
    CA11006-400
  • Supplier No.:
    CC-3171
Specifications
About this item
BEBM® is a basal medium sold separately. This is used as a component in the BEGM® Bronchial Epithelial Cell Growth Medium BulletKit® (catalog number CC-3170) which consists of basal medium and supplements packaged together.
BEGM® Bronchial Epithelial Cell Growth Mediumis specially designed to support the growth of human primary bronchial epithelial cells. BEGM® BulletKit® is serum-free and has been optimized to grow cells from normal and diseased tissues.
Published research shows BEGM® BulletKit® has expanded applications. Our customers have used BEGM® BulletKit® to culture well-established cell lines such as BEAS-2B, home-grown bronchial epithelial cells and supporting co-culture models with breast cancer cell lines such as MCF-7 and MDA-MB-231.
BEGM® is an optimized and readily available media kit providing researchers the confidence and the convenience of a system that works to support their cells. This is especially important for researchers who are utilizing cell lines as a stepping stone to primary cell culture. Working with the same BEGM® Media between cell lines and primary NHBE can reduce variability within the experiments and simplify the shift for researchers. In addition, if primary cells are utilized as a side-by-side control with normal cell lines or home-grown bronchial epithelial cells, the BEGM® BulletKit® Media may also offer cost-benefit since customers can use the same media to support multiple cell types.
Delivery: Content: 1 × Bottle Basal Medium 500 ml.