PrEBM® Prostate Epithelial Cell Growth Basal Medium
CA10065-272
:
- Cat. no.:CA10065-272
- Supplier No.:CC-3165
PrEBM® is a prostate epithelial cell growth basal medium sold separately. This can be used with 1 kit of CC-4177 (prostate epithelial cell growth medium SingleQuots® Supplements and Growth Factors). The full PrEGM® BulletKit® consisting of basal medium and supplements can be purchased together using the catalog number CC-3166.
PrEGM® Bulletkit®, Prostate Epithelial Cell Growth Medium has been specially formulated for the maintenance and expansion of human prostate epithelial cells. PrEGM® Medium contains no serum.
: Content: 1 × Bottle Basal Medium 500 ml